4-Amino-2,6-dimethylpyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2091-25G A2091-5G
TCA2091-25GEA 222.78 CAD
TCA2091-25G TCA2091-5G
4-Amino-2,6-dimethylpyrimidine ≥98.0% (by GC, titration analysis)
4-Amino-2,6-dimethylpyrimidine

CAS Number: 461-98-3
MDL Number: MFCD00006106
Molecular Formula: C6H9N3
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 254
Melting point (°C): 185

Formula: C₆H₉N₃
Boiling Pt: 254 °C
Melting Pt: 185 °C
MDL Number: MFCD00006106
CAS Number: 461-98-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f4473fc32857276f045cc5e4ece46ef26166d65&vid=1f4473fc32857276f045cc5e4ece46ef26166d65&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR