N4-Acetylcytosine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A2089-25G
A2089-5G
TCA2089-25GEA
143.78
CAD
TCA2089-25G
TCA2089-5G
N4-Acetylcytosine ≥98.0% (by titrimetric analysis)
N4-Acetylcytosine
CAS Number: 14631-20-0
MDL Number: MFCD00134466
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 325
Formula:
C₆H₇N₃O₂ Melting Pt: 325 °C |
MDL Number:
MFCD00134466 CAS Number: 14631-20-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...