2-Amino-3-benzyloxypyridine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A2080-5G
A2080-25G
TCA2080-5GEA
56.88
CAD
TCA2080-5G
TCA2080-25G
2-Amino-3-benzyloxypyridine ≥98.0% (by titrimetric analysis)
2-Amino-3-benzyloxypyridine
CAS Number: 24016-03-3
MDL Number: MFCD00006316
Molecular Formula: C12H12N2O
Molecular Weight: 200.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 97
Formula:
C₁₂H₁₂N₂O Melting Pt: 97 °C |
MDL Number:
MFCD00006316 CAS Number: 24016-03-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...