5-Aminoindole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2071-1G A2071-5G
TCA2071-1GEA 96.38 CAD
TCA2071-1G TCA2071-5G
5-Aminoindole ≥98.0% (by GC, titration analysis)
5-Aminoindole

CAS Number: 5192-03-0
MDL Number: MFCD00005679
Molecular Formula: C8H8N2
Molecular Weight: 132.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 190
Melting point (°C): 133

Formula: C₈H₈N₂
Boiling Pt: 190 °C
Melting Pt: 133 °C
MDL Number: MFCD00005679
CAS Number: 5192-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13b88655704a2d298fbdd17065aef469c16f1f86&vid=13b88655704a2d298fbdd17065aef469c16f1f86&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR