2-Amino-4,5-dimethylthiazole hydrobromide ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-4,5-dimethylthiazole Hydrobromide, 4,5-Dimethylthiazol-2-amine hydrobromide

A2062-25G A2062-5G
TCA2062-25GEA 278.08 CAD
TCA2062-25G TCA2062-5G
2-Amino-4,5-dimethylthiazole hydrobromide ≥98.0%
2-Amino-4,5-dimethylthiazole hydrobromide

CAS Number: 7170-76-5
MDL Number: MFCD00035212
Molecular Formula: C5H8N2S
Molecular Weight: 209.11
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal

Formula: C₅H₉BrN₂S
MW: 209.11 g/mol
MDL Number: MFCD00035212
CAS Number: 7170-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09eabda551cd4ba06de987ae6b108dd131d13eaa&vid=09eabda551cd4ba06de987ae6b108dd131d13eaa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR