2-Acetyl-6-methoxynaphthalene ≥98.0%

Supplier: TCI America

A2030-25G A2030-500G A2030-100G
TCA2030-25GEA 110.6 CAD
TCA2030-25G TCA2030-500G TCA2030-100G
2-Acetyl-6-methoxynaphthalene ≥98.0%
2-Acetyl-6-methoxynaphthalene

CAS Number: 3900-45-6
MDL Number: MFCD00021643
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 195
Melting point (°C): 109


Other Product Names / Synonyms: 6'-Methoxy-2'-acetonaphthone

Formula: C₁₃H₁₂O₂
Boiling Pt: 195 °C
Melting Pt: 109 °C
MDL Number: MFCD00021643
CAS Number: 3900-45-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32fe29e34a5fed928a3641af60178656f4c591dc&vid=32fe29e34a5fed928a3641af60178656f4c591dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR