L-Arginine methyl ester dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

A2017-25G A2017-5G
TCA2017-25GEA 151.13 CAD
TCA2017-25G TCA2017-5G
L-Arginine methyl ester dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
L-Arginine methyl ester dihydrochloride

CAS Number: 26340-89-6
MDL Number: MFCD00038948
Molecular Formula: C7H16N4O2
Molecular Weight: 261.15
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 193
Specific rotation [a]20/D: 21 deg (C=2.5, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Arg-OMe.2HCl

Formula: C₇H₁₆N₄O₂
Melting Pt: 193 °C
MDL Number: MFCD00038948
CAS Number: 26340-89-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dbcf0ff06748d981758b3642dde0681b2a3ff90&vid=2dbcf0ff06748d981758b3642dde0681b2a3ff90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR