2-Acetyl-3-ethylpyrazine ≥98.0%
Supplier: TCI America
A2003-1G
A2003-5G
TCA2003-1GEA
45.82
CAD
TCA2003-1G
TCA2003-5G
2-Acetyl-3-ethylpyrazine ≥98.0%
2-Acetyl-3-ethylpyrazine
CAS Number: 32974-92-8
MDL Number: MFCD00038028
Molecular Formula: C8H10N2O
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 77
Flash Point (°C): 87
Specific Gravity (20/20): 1.08
Formula:
C₈H₁₀N₂O Boiling Pt: 77 °C Density: 1.08 (20 °C) Flash Pt: 87 °C |
MDL Number:
MFCD00038028 CAS Number: 32974-92-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...