2-Amino-5-chlorobenzamide ≥98.0%
Supplier: TCI America
A1970-1G
A1970-5G
TCA1970-1GEA
39.5
CAD
TCA1970-1G
TCA1970-5G
2-Amino-5-chlorobenzamide ≥98.0%
2-Amino-5-chlorobenzamide
CAS Number: 5202-85-7
MDL Number: MFCD00017126
Molecular Formula: C7H7ClN2O
Molecular Weight: 170.60
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 171
Other Product Names / Synonyms: 5-Chloroanthranilamide
Formula:
C₇H₇ClN₂O Melting Pt: 171 °C |
MDL Number:
MFCD00017126 CAS Number: 5202-85-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...