2-Amino-5-(trifluoromethyl)pyridine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
5-(Trifluoromethyl)pyridin-2-amine, 5-(Trifluoromethyl)-2-pyridylamine
A1968-25G
A1968-5G
TCA1968-25GEA
356.91
CAD
TCA1968-25G
TCA1968-5G
2-Amino-5-(trifluoromethyl)pyridine ≥97.0% (by GC, titration analysis)
2-Amino-5-(trifluoromethyl)pyridine
CAS Number: 74784-70-6
MDL Number: MFCD00042164
Molecular Formula: C6H5F3N2
Molecular Weight: 162.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 47
Formula:
C₆H₅F₃N₂ MW: 162.11 Melting Pt: 47 °C |
MDL Number:
MFCD00042164 CAS Number: 74784-70-6 UN: 2811 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...