5-Amino-2-methoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1956-25G A1956-5G
TCA1956-25GEA 276.03 CAD
TCA1956-25G TCA1956-5G
5-Amino-2-methoxypyridine ≥98.0% (by GC, titration analysis)
5-Amino-2-methoxypyridine

CAS Number: 6628-77-9
MDL Number: MFCD00006264
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 113
Specific Gravity (20/20): 1.17
Storage Temperature: 0-10°C

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 120 °C
Density: 1.17 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00006264
CAS Number: 6628-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=554da6a715511426d6ac0822be7e55a4e8d83530&vid=554da6a715511426d6ac0822be7e55a4e8d83530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR