3-Amino-p-anisic acid ≥98.0%

Supplier: TCI America

A1955-25G A1955-5G
TCA1955-25GEA 671.5 CAD
TCA1955-25G TCA1955-5G
3-Amino-p-anisic acid ≥98.0%
3-Amino-p-anisic acid

CAS Number: 2840-26-8
MDL Number: MFCD00002521
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 205


Other Product Names / Synonyms: 3-Amino-p-anisic Acid

Formula: C₈H₉NO₃
Melting Pt: 205 °C
MDL Number: MFCD00002521
CAS Number: 2840-26-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed767650c4c85b28cadc6e74c1cc5928ba748d0b&vid=ed767650c4c85b28cadc6e74c1cc5928ba748d0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR