3-Acetylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
A1942-5G
A1942-25G
TCA1942-5GEA
140.62
CAD
TCA1942-5G
TCA1942-25G
3-Acetylphenylboronic acid (contains varying amounts of Anhydride)
3-Acetylphenylboronic acid
CAS Number: 204841-19-0
MDL Number: MFCD01074678
Molecular Formula: C8H9BO3
Molecular Weight: 163.97
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 208
Other Product Names / Synonyms: 3-Acetylbenzeneboronic Acid
Formula:
C₈H₉BO₃ Melting Pt: 208 °C |
MDL Number:
MFCD01074678 CAS Number: 204841-19-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...