3-Amino-2-cyclohexen-1-one ≥98.0%

Supplier: TCI America

A1936-25G A1936-5G
TCA1936-25GEA 215.43 CAD
TCA1936-25G TCA1936-5G
3-Amino-2-cyclohexen-1-one ≥98.0%
3-Amino-2-cyclohexen-1-one

CAS Number: 5220-49-5
MDL Number: MFCD00013783
Molecular Formula: C6H9NO
Molecular Weight: 111.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 132

Formula: C₆H₉NO
Melting Pt: 132 °C
MDL Number: MFCD00013783
CAS Number: 5220-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=251f0d04f36a6419901dc4215004ebb7fc49de3b&vid=251f0d04f36a6419901dc4215004ebb7fc49de3b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR