3-Bromo-5-(trifluoromethyl)aniline ≥98.0%

Supplier: TCI America

A1914-5G A1914-25G
TCA1914-5GEA 71.1 CAD
TCA1914-5G TCA1914-25G
3-Bromo-5-(trifluoromethyl)aniline ≥98.0%
3-Bromo-5-(trifluoromethyl)aniline

CAS Number: 54962-75-3
Molecular Formula: C7H5BrF3N
Molecular Weight: 240.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 223
Flash Point (°C): 107
Specific Gravity (20/20): 1.72


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Bromo-5-(trifluoromethyl)aniline

Formula: C₇H₅BrF₃N
Boiling Pt: 223 °C
Density: 1.72 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00236205
CAS Number: 54962-75-3
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b30d8bc2a2788e79169d5a16f7bc150ef5196930&vid=9adbe49dfb87450bcdfb1c2246692ff5349ec0c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR