4-Acetylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane

A1907-1G A1907-5G
TCA1907-1GEA 33.18 CAD
TCA1907-1G TCA1907-5G
4-Acetylphenylboronic acid (contains varying amounts of Anhydride)
4-Acetylphenylboronic acid

CAS Number: 149104-90-5
Molecular Formula: C8H9BO3
Molecular Weight: 163.97
Form: Crystal
Color: White
Melting point (°C): 239


Other Product Names / Synonyms: 4-Acetylbenzeneboronic Acid

Formula: C₈H₉BO₃
Melting Pt: 239 °C
MDL Number: MFCD01074667
CAS Number: 149104-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=866742797b72826eadd66e576f0e5510cad6bb4b&vid=866742797b72826eadd66e576f0e5510cad6bb4b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR