4-Amino-5-chloro-2,6-dimethylpyrimidine ≥98.0%

Supplier: TCI America

A1901-25G A1901-5G
TCA1901-25GEA 366.56 CAD
TCA1901-25G TCA1901-5G
4-Amino-5-chloro-2,6-dimethylpyrimidine ≥98.0%
4-Amino-5-chloro-2,6-dimethylpyrimidine

CAS Number: 2858-20-0
Molecular Formula: C6H8ClN3
Molecular Weight: 157.60
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 165

Formula: C₆H₈ClN₃
MW: 157.60 g/mol
Melting Pt: 165 °C
MDL Number: MFCD00191727
CAS Number: 2858-20-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87094236b0da10b061de201b90db282a66f4f307&vid=87094236b0da10b061de201b90db282a66f4f307&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR