3-Amino-2-bromopyridine ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromopyridin-3-amine, 2-Bromo-3-pyridylamine

A1898-5G A1898-1G
TCA1898-5GEA 168.59 CAD
TCA1898-5G TCA1898-1G
3-Amino-2-bromopyridine ≥98.0%
3-Amino-2-bromopyridine

CAS Number: 39856-58-1
Molecular Formula: C5H5BrN2
Molecular Weight: 173.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 79

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00234064
CAS Number: 39856-58-1
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=935f36be262e42944c96bbbd59fdfa89a31ad10e&vid=935f36be262e42944c96bbbd59fdfa89a31ad10e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR