2-Amino-5-bromo-4-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 5-Bromo-4-methylpyridin-2-amine, 5-Bromo-4-methyl-2-pyridylamine, 2-Amino-5-bromo-4-picoline

A1897-1G A1897-5G
TCA1897-1GEA 26.86 CAD
TCA1897-1G TCA1897-5G
2-Amino-5-bromo-4-methylpyridine ≥98.0%
2-Amino-5-bromo-4-methylpyridine

CAS Number: 98198-48-2
Molecular Formula: C6H7BrN2
Molecular Weight: 187.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 151


Other Product Names / Synonyms: 2-Amino-5-bromo-4-picoline

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 151 °C
MDL Number: MFCD03427660
CAS Number: 98198-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dddd60303b0ffe097bc83bd1315dd76f20f275c9&vid=dddd60303b0ffe097bc83bd1315dd76f20f275c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR