2-Amino-5-nitro-3-picoline ≥98.0%
Supplier: TCI America
Synonyms:
2-Amino-3-methyl-5-nitropyridine, 3-Methyl-5-nitro-2-pyridylamine
A1891-5G
A1891-1G
TCA1891-5GEA
120.08
CAD
TCA1891-5G
TCA1891-1G
2-Amino-5-nitro-3-picoline ≥98.0%
2-Amino-5-nitro-3-picoline
CAS Number: 18344-51-9
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 261
Melting point (°C): 255
Other Product Names / Synonyms: 2-Amino-5-nitro-3-picoline
Formula:
C₆H₇N₃O₂ Boiling Pt: 261 °C Melting Pt: 255 °C |
MDL Number:
MFCD03095060 CAS Number: 18344-51-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...