5-Amino-2-bromopyridine ≥97.0%

Supplier: TCI America

A1890-5G A1890-1G
TCA1890-5GEA 172.22 CAD
TCA1890-5G TCA1890-1G
5-Amino-2-bromopyridine ≥97.0%
5-Amino-2-bromopyridine

CAS Number: 13534-97-9
MDL Number: MFCD01646116
Molecular Formula: C5H5BrN2
Molecular Weight: 173.01
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 77

Formula: C₅H₅BrN₂
Boiling Pt: 180 °C
Melting Pt: 77 °C
MDL Number: MFCD01646116
CAS Number: 13534-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9f7e9d4a3e7f8972bd88fd1b4e77c959299482f&vid=e9f7e9d4a3e7f8972bd88fd1b4e77c959299482f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR