6-Amino-3-bromo-2-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 6-Amino-3-bromo-2-picoline, 6-AMINO-3-BROMO-2-METHYLPYRIDINE, 3-Bromo-2-methyl-6-pyridylamine, 5-Bromo-6-methylpyridin-2-amine, 3-Bromo-2-methylpyridin-6-amine, 2-Amino-5-bromo-6-picoline, ['6-Amino-3-bromo-2-picoline'], 5-Bromo-6-methyl-2-pyridylamine, 2-Amino-5-bromo-6-methylpyridine

A1889-1G A1889-5G
TCA1889-1GEA 28.44 CAD
TCA1889-1G TCA1889-5G
6-Amino-3-bromo-2-methylpyridine ≥98.0%
6-Amino-3-bromo-2-methylpyridine

CAS Number: 42753-71-9
Molecular Formula: C6H7BrN2
Molecular Weight: 187.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 83


Other Product Names / Synonyms: 6-Amino-3-bromo-2-picoline

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 83 °C
MDL Number: MFCD00068230
CAS Number: 42753-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21c0d386ff3b8e3f14386476b7cb78710ddf77fa&vid=21c0d386ff3b8e3f14386476b7cb78710ddf77fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR