2-Amino-4-methyl-3-nitropyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1888-1G A1888-5G
TCA1888-1GEA 39.5 CAD
TCA1888-1G TCA1888-5G
2-Amino-4-methyl-3-nitropyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-methyl-3-nitropyridine

CAS Number: 6635-86-5
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138


Other Product Names / Synonyms: 2-Amino-3-nitro-4-picoline

Formula: C₆H₇N₃O₂
Melting Pt: 138 °C
MDL Number: MFCD00006315
CAS Number: 6635-86-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=963294086640316bd8a62b0869f5ca183bedcee8&vid=963294086640316bd8a62b0869f5ca183bedcee8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR