3-Amino-2-chloro-4-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Amino-2-chloro-4-methylpyridine, 3-Amino-2-chloro-4-picoline, 2-Chloro-4-methylpyridin-3-amine

A1855-5G A1855-25G
TCA1855-5GEA 63.2 CAD
TCA1855-5G TCA1855-25G
3-Amino-2-chloro-4-methylpyridine ≥98.0% (by GC, titration analysis)
3-Amino-2-chloro-4-methylpyridine

CAS Number: 133627-45-9
Molecular Formula: C6H7ClN2
Molecular Weight: 142.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 68


Other Product Names / Synonyms: 3-Amino-2-chloro-4-picoline

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Melting Pt: 68 °C
MDL Number: MFCD00673152
CAS Number: 133627-45-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f2f9582d01e61b09e9d145d238c5a70f5217100&vid=2f2f9582d01e61b09e9d145d238c5a70f5217100&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR