4-Bromo-3-(trifluoromethyl)aniline ≥98.0%
Supplier: TCI America
Synonyms:
5-Amino-2-bromobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-m-toluidine, 5-Amino-2-bromo-α,α,α-trifluorotoluene
CAS Number: 393-36-2
Molecular Formula: C7H5BrF3N
Molecular Weight: 240.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 108
Melting point (°C): 50
Flash Point (°C): 113
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Amino-1-bromo-2-(trifluoromethyl)benzene, 4-Bromo-3-(trifluoromethyl)aniline
Formula:
C₇H₅BrF₃N Boiling Pt: 108 °C Melting Pt: 50 °C Flash Pt: 113 °C |
MDL Number:
MFCD00007827 CAS Number: 393-36-2 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...