1-Acetyl-4-(4-hydroxyphenyl)piperazine ≥98.0%

Supplier: TCI America

Synonyms: 1-(4-(4-Hydroxyphenyl)piperazin-1-yl)ethanone, 4-(4-Acetyl-1-piperazinyl)phenol

A1845-25G A1845-250G
TCA1845-25GEA 36.34 CAD
TCA1845-25G TCA1845-250G
1-Acetyl-4-(4-hydroxyphenyl)piperazine ≥98.0%
1-Acetyl-4-(4-hydroxyphenyl)piperazine

CAS Number: 67914-60-7
Molecular Formula: C12H16N2O2
Molecular Weight: 220.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 185


Other Product Names / Synonyms: 4-(4-Acetyl-1-piperazinyl)phenol

Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00044905
CAS Number: 67914-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c386da08f5c914282456d4a6601ca5686af76d6d&vid=c386da08f5c914282456d4a6601ca5686af76d6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR