2-Amino-5-iodopyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1842-25G A1842-1G A1842-5G
TCA1842-25GEA 386.78 CAD
TCA1842-25G TCA1842-1G TCA1842-5G
2-Amino-5-iodopyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-iodopyridine

CAS Number: 20511-12-0
Molecular Formula: C5H5IN2
Molecular Weight: 220.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 127
Storage Temperature: 0-10°C

Formula: C₅H₅IN₂
Melting Pt: 127 °C
MDL Number: MFCD00160312
CAS Number: 20511-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8496f989134c7767c6d40adb89cadbc59d0a64ad&vid=8496f989134c7767c6d40adb89cadbc59d0a64ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR