2-Amino-3-formylpyridine ≥97.0%

Supplier: TCI America

Synonyms: 2-Amino-3-pyridinecarboxaldehyde, 2-Aminonicotinaldehyde, 2-Aminopyridine-3-carboxaldehyde

A1818-5G A1818-1G
TCA1818-5GEA 244.9 CAD
TCA1818-5G TCA1818-1G
2-Amino-3-formylpyridine ≥97.0%
2-Amino-3-formylpyridine

CAS Number: 7521-41-7
Molecular Formula: C6H6N2O
Molecular Weight: 122.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 98
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Aminopyridine-3-carboxaldehyde

Formula: C₆H₆N₂O
MW: 122.13
Melting Pt: 98 °C
MDL Number: MFCD01830382
CAS Number: 7521-41-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c3825cbd52052709688ef04d89d672443c857f9&vid=9c3825cbd52052709688ef04d89d672443c857f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR