3-Amino-6-chloropyridazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1817-25G A1817-5G
TCA1817-25GEA 276.5 CAD
TCA1817-25G TCA1817-5G
3-Amino-6-chloropyridazine ≥98.0% (by GC, titration analysis)
3-Amino-6-chloropyridazine

CAS Number: 5469-69-2
Molecular Formula: C4H4ClN3
Molecular Weight: 129.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₄H₄ClN₃
MDL Number: MFCD00051506
CAS Number: 5469-69-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdba5cb0685f245c2dffe0968ab4b905d39b4db2&vid=fdba5cb0685f245c2dffe0968ab4b905d39b4db2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR