2-Amino-3,5-dibromopyrazine ≥98.0%

Supplier: TCI America

Synonyms: Amino-3,5-dibromopyrazine

A1806-5G A1806-1G
TCA1806-5GEA 131.14 CAD
TCA1806-5G TCA1806-1G
2-Amino-3,5-dibromopyrazine ≥98.0%
2-Amino-3,5-dibromopyrazine

CAS Number: 24241-18-7
Molecular Formula: C4H3Br2N3
Molecular Weight: 252.90
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 114

Formula: C₄H₃Br₂N₃
MW: 252.90 g/mol
Melting Pt: 114 °C
MDL Number: MFCD00673150
CAS Number: 24241-18-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac9d4d421d50f2198c45e02d52d38a879f741489&vid=ac9d4d421d50f2198c45e02d52d38a879f741489&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR