4-Acetamidobenzenesulfonyl azide ≥98.0%

Supplier: TCI America

A1786-25G A1786-5G A1786-100G
TCA1786-25GEA 184.77 CAD
TCA1786-25G TCA1786-5G TCA1786-100G
4-Acetamidobenzenesulfonyl azide ≥98.0%
4-Acetamidobenzenesulphonyl azide

CAS Number: 2158-14-7
MDL Number: MFCD00029626
Molecular Formula: C8H8N4O3S
Molecular Weight: 240.24
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 112

Formula: C₈H₈N₄O₃S
MW: 240.24 g/mol
Melting Pt: 112 °C
MDL Number: MFCD00029626
CAS Number: 2158-14-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f2a2fc0540dc4e562bb67f6d700c9e222d87a32&vid=4f2a2fc0540dc4e562bb67f6d700c9e222d87a32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR