2-Amino-5-bromopyrazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1683-5G A1683-1G
TCA1683-5GEA 221.52 CAD
TCA1683-5G TCA1683-1G
2-Amino-5-bromopyrazine ≥98.0% (by GC, titration analysis)
2-Amino-5-bromopyrazine

CAS Number: 59489-71-3
Molecular Formula: C4H4BrN3
Molecular Weight: 174.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 113

Formula: C₄H₄BrN₃
Melting Pt: 113 °C
MDL Number: MFCD00235015
CAS Number: 59489-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0230bb8dcd59e5d9c343da9cb0746588c69a1e2f&vid=0230bb8dcd59e5d9c343da9cb0746588c69a1e2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR