2-Aminoimidazole hemisulfate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A1665-1G
A1665-5G
TCA1665-1GEA
41.08
CAD
TCA1665-1G
TCA1665-5G
2-Aminoimidazole hemisulfate ≥98.0% (by HPLC, titration analysis)
2-Aminoimidazole hemisulphate
CAS Number: 1450-93-7
Molecular Formula: C6H10N6
Molecular Weight: 264.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 270
Other Product Names / Synonyms: Bis(2-aminoimidazole) Sulfate
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...