2-Amino-5-fluoropyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1664-1G A1664-5G
TCA1664-1GEA 85.95 CAD
TCA1664-1G TCA1664-5G
2-Amino-5-fluoropyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-fluoropyridine

CAS Number: 21717-96-4
Molecular Formula: C5H5FN2
Molecular Weight: 112.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 97

Formula: C₅H₅FN₂
MW: 112.11
Melting Pt: 97 °C
MDL Number: MFCD01861120
CAS Number: 21717-96-4
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1363fe35f6160a3709837f78780a74468ce034bd&vid=1363fe35f6160a3709837f78780a74468ce034bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR