trans-4-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1656-1G A1656-5G A1656-25G
TCA1656-1GEA 107.44 CAD
TCA1656-1G TCA1656-5G TCA1656-25G
trans-4-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-Aminocyclohexanecarboxylic acid

CAS Number: 3685-25-4
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 495

Formula: C₇H₁₃NO₂
Melting Pt: 495 °C
MDL Number: MFCD00209953
CAS Number: 3685-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2fa7f18d4828427b093e2f717ae6532ff3e4cb7&vid=e2fa7f18d4828427b093e2f717ae6532ff3e4cb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR