4-Acetamidopiperidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A1653-5G
A1653-25G
TCA1653-5GEA
173.48
CAD
TCA1653-5G
TCA1653-25G
4-Acetamidopiperidine ≥98.0% (by GC, titration analysis)
4-Acetamidopiperidine
CAS Number: 5810-56-0
MDL Number: MFCD05254805
Molecular Formula: C7H14N2O
Molecular Weight: 142.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 141
Formula:
C₇H₁₄N₂O Melting Pt: 141 °C |
MDL Number:
MFCD05254805 CAS Number: 5810-56-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...