3-Acetylbenzoic acid ≥97.0%
Supplier: TCI America
Synonyms:
3'-Acetophenonecarboxylic acid, 3-Acetylbenzoic Acid
A1652-1G
A1652-5G
TCA1652-1GEA
372.88
CAD
TCA1652-1G
TCA1652-5G
3-Acetylbenzoic acid ≥97.0%
3-Acetylbenzoic acid
CAS Number: 586-42-5
MDL Number: MFCD00045847
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 164
Other Product Names / Synonyms: 3'-Acetophenonecarboxylic Acid
Formula:
C₉H₈O₃ Melting Pt: 164 °C |
MDL Number:
MFCD00045847 CAS Number: 586-42-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...