4-(Aminomethyl)benzonitrile hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 4-(Aminomethyl)benzonitrile HCl, α-Amino-p-tolunitrile Hydrochloride, 4-(Aminomethyl)benzonitrile Hydrochloride, 4-Cyanobenzylamine Hydrochloride

A1645-1G A1645-5G
TCA1645-1GEA 52.14 CAD
TCA1645-1G TCA1645-5G
4-(Aminomethyl)benzonitrile hydrochloride ≥98.0%
4-(Aminomethyl)benzonitrile hydrochloride

CAS Number: 15996-76-6
MDL Number: MFCD01861472
Molecular Formula: C8H8N2
Molecular Weight: 168.62
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: alpha-Amino-p-tolunitrile Hydrochloride, 4-Cyanobenzylamine Hydrochloride

Formula: C₈H₉ClN₂
MW: 168.62 g/mol
MDL Number: MFCD01861472
CAS Number: 15996-76-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aa5c1093148befc42ec603f5c3735fc28d8471c&vid=5aa5c1093148befc42ec603f5c3735fc28d8471c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR