2-Amino-5-nitro-4-picoline ≥98.0%

Supplier: TCI America

Synonyms: 4-Methyl-5-nitro-2-pyridylamine, 4-Methyl-5-nitropyridin-2-amine, 2-Amino-4-methyl-5-nitropyridine

A1638-1G A1638-5G
TCA1638-1GEA 52.93 CAD
TCA1638-1G TCA1638-5G
2-Amino-5-nitro-4-picoline ≥98.0%
2-Amino-5-nitro-4-picoline

CAS Number: 21901-40-6
MDL Number: MFCD00010692
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 224


Other Product Names / Synonyms: 2-Amino-5-nitro-4-picoline

Formula: C₆H₇N₃O₂
MW: 153.14
Melting Pt: 220
MDL Number: MFCD00010692
CAS Number: 21901-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0652f2f90cf9251386e4b554b1829fa022496808&vid=0652f2f90cf9251386e4b554b1829fa022496808&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR