1-Adamantaneethanol ≥98.0%

Supplier: TCI America

Synonyms: 1-(2-Hydroxyethyl)adamantane, 1-(2-Hydroxyethyl)tricyclo[3.3.1.1(3,7)]decane, 2-(1-Adamantyl)ethanol

A1619-25G A1619-5G
TCA1619-25GEA 649.38 CAD
TCA1619-25G TCA1619-5G
1-Adamantaneethanol ≥98.0%
1-Adamantaneethanol

CAS Number: 6240-11-5
MDL Number: MFCD00074756
Molecular Formula: C12H20O
Molecular Weight: 180.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 76


Other Product Names / Synonyms: 1-(2-Hydroxyethyl)adamantane

Formula: C₁₂H₂₀O
MW: 180.29 g/mol
Melting Pt: 76 °C
MDL Number: MFCD00074756
CAS Number: 6240-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d3eea10a273dfc04c25f8027e929e893e2737af&vid=9d3eea10a273dfc04c25f8027e929e893e2737af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR