Ethyl-4-amino-1-piperidinecarboxylate ≥97.0%
Supplier: TCI America
A1595-25G
A1595-5G
TCA1595-25GEA
126.4
CAD
TCA1595-25G
TCA1595-5G
Ethyl-4-amino-1-piperidinecarboxylate ≥97.0%
Ethyl-4-amino-1-piperidinecarboxylate
CAS Number: 58859-46-4
MDL Number: MFCD00006484
Molecular Formula: C8H16N2O2
Molecular Weight: 172.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 78
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: 4-Amino-1-piperidinecarboxylic Acid Ethyl Ester
Formula:
C₈H₁₆N₂O₂ Density: 1.08 (20 °C) Flash Pt: 78 °C |
MDL Number:
MFCD00006484 CAS Number: 58859-46-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...