3-Amino-4-pyrazolecarbonitrile ≥98.0%

Supplier: TCI America

Synonyms: 5-Aminopyrazole-4-carbonitrile, 3-Amino-1H-pyrazole-4-carbonitrile, 3-Aminopyrazole-4-carbonitrile, 3-Amino-4-cyanopyrazole 3-Amino-4-pyrazolecarbonitrile 5-Amino-4-cyanopyrazole 3-Amino-1H-pyrazole-4-carbonitrile, 3-Amino-4-cyanopyrazole

A1594-25G A1594-5G
TCA1594-25GEA 252.42 CAD
TCA1594-25G TCA1594-5G
3-Amino-4-pyrazolecarbonitrile ≥98.0%
3-Amino-4-pyrazolecarbonitrile

CAS Number: 16617-46-2
MDL Number: MFCD00005237
Molecular Formula: C4H4N4
Molecular Weight: 108.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 175


Other Product Names / Synonyms: 3-Amino-4-cyanopyrazole

Formula: C₄H₄N₄
MW: 108.10 g/mol
Melting Pt: 175 °C
MDL Number: MFCD00005237
CAS Number: 16617-46-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5e5d66a7c4a48b37c76fb3305aace626207ee6c&vid=a5e5d66a7c4a48b37c76fb3305aace626207ee6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR