3-Amino-2-naphthol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Amino-2-hydroxynaphthalene, 3-Aminonaphthalen-2-ol

A1593-5G A1593-1G
TCA1593-5GEA 638 CAD
TCA1593-5G TCA1593-1G
3-Amino-2-naphthol ≥98.0% (by GC, titration analysis)
3-Amino-2-naphthol

CAS Number: 5417-63-0
MDL Number: MFCD00004113
Molecular Formula: C10H9NO
Molecular Weight: 159.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD00004113
CAS Number: 5417-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caace4187ede00c1e56626725ae55e49e4f71072&vid=caace4187ede00c1e56626725ae55e49e4f71072&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR