3-Amino-5-hydroxy-1H-pyrazole ≥98.0%

Supplier: TCI America

Synonyms: 3-Amino-5-hydroxy-1H-pyrazole, 3-Amino-5-hydroxypyrazole, 5-Amino-3-pyrazolol, 3-Amino-1H-pyrazol-5(4H)-one, 3-Amino-2-pyrazolin-5-one, 5-Hydroxy-3-aminopyrazole, 5-Amino-2,4-dihydro-3H-pyrazol-3-one

A1589-5G A1589-25G
TCA1589-5GEA 135.56 CAD
TCA1589-5G TCA1589-25G
3-Amino-5-hydroxy-1H-pyrazole ≥98.0%
3-Amino-5-hydroxy-1H-pyrazole

CAS Number: 6126-22-3
MDL Number: MFCD00022384
Molecular Formula: C3H5N3O
Molecular Weight: 99.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 5-Amino-2,4-dihydro-3H-pyrazol-3-one, 5-Amino-3-pyrazolol, 5-Hydroxy-3-aminopyrazole

Formula: C₃H₅N₃O
MDL Number: MFCD00022384
CAS Number: 6126-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87ecafdde0659e7027464508989ba718a8d36bc4&vid=87ecafdde0659e7027464508989ba718a8d36bc4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR