2-Amino-3,5-dibromopyridine ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Dibromo-2-pyridylamine, 3,5-Dibromopyridin-2-amine

A1588-25G A1588-5G
TCA1588-25GEA 45.82 CAD
TCA1588-25G TCA1588-5G
2-Amino-3,5-dibromopyridine ≥98.0%
2-Amino-3,5-dibromopyridine

CAS Number: 35486-42-1
MDL Number: MFCD00038041
Molecular Formula: C5H4Br2N2
Molecular Weight: 251.91
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 105

Formula: C₅H₄Br₂N₂
MW: 251.91 g/mol
Boiling Pt: 254
Melting Pt: 104
MDL Number: MFCD00038041
CAS Number: 35486-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a2d8cd56b14134778d12618641d161916c67c46&vid=9a2d8cd56b14134778d12618641d161916c67c46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR