2-Fluoro-5-(trifluoromethyl)aniline ≥98.0%
Supplier: TCI America
Synonyms:
3-Amino-alpha,alpha,alpha,4-tetrafluorotoluene, α,α,α,6-Tetrafluoro-m-toluidine, 3-Amino-α,α,α,4-tetrafluorotoluene, 3-Amino-4-fluorobenzotrifluoride
CAS Number: 535-52-4
MDL Number: MFCD00007653
Molecular Formula: C7H5F4N
Molecular Weight: 179.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 70
Specific Gravity (20/20): 1.40
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-5-trifluoromethylaniline
Formula:
C₇H₅F₄N MW: 179.12 g/mol Boiling Pt: 190 °C Density: 1.40 (20 °C) Flash Pt: 70 °C |
MDL Number:
MFCD00007653 CAS Number: 535-52-4 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...