2-Amino-5-bromothiazole hydrobromide ≥95.0%

Supplier: TCI America

Synonyms: 2-Amino-5-bromothiazole monohydrobromide, 2-Amino-5-bromothiazole HBr

A1573-25G A1573-5G
TCA1573-25GEA 252.8 CAD
TCA1573-25G TCA1573-5G
2-Amino-5-bromothiazole hydrobromide ≥95.0%
2-Amino-5-bromothiazole hydrobromide

CAS Number: 61296-22-8
MDL Number: MFCD00012712
Molecular Formula: C3H3BrN2S
Molecular Weight: 259.95
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 163

Formula: C₃H₃BrN₂S
Melting Pt: 163 °C
MDL Number: MFCD00012712
CAS Number: 61296-22-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a9e7bd801df3196e4104a4404e8fb16c60dff41&vid=9a9e7bd801df3196e4104a4404e8fb16c60dff41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR