3-Acetoxy-2-methylbenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A1558-5G A1558-25G
TCA1558-5GEA 52.93 CAD
TCA1558-5G TCA1558-25G
3-Acetoxy-2-methylbenzoic acid ≥98.0% (by HPLC, titration analysis)
3-Acetoxy-2-methylbenzoic acid

CAS Number: 168899-58-9
MDL Number: MFCD00957176
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 148


Other Product Names / Synonyms: 3-Acetoxy-o-toluic Acid

Formula: C₁₀H₁₀O₄
Melting Pt: 148 °C
MDL Number: MFCD00957176
CAS Number: 168899-58-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81e5df5f175fbf31d50a69684d2c7b2c5214022c&vid=81e5df5f175fbf31d50a69684d2c7b2c5214022c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR