2-Amino-5-chloro-3-nitropyridine ≥98.0%

Supplier: TCI America

Synonyms: 5-Chloro-3-nitro-2-pyridylamine, 5-Chloro-3-nitropyridin-2-amine

A1557-25G A1557-5G
TCA1557-25GEA 186.44 CAD
TCA1557-25G TCA1557-5G
2-Amino-5-chloro-3-nitropyridine ≥98.0%
2-Amino-5-chloro-3-nitropyridine

CAS Number: 5409-39-2
MDL Number: MFCD00092011
Molecular Formula: C5H4ClN3O2
Molecular Weight: 173.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 196

Formula: C₅H₄ClN₃O₂
Melting Pt: 196 °C
MDL Number: MFCD00092011
CAS Number: 5409-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ade75f1ca691aadc9841f9111bd9a7ca32550a7&vid=5ade75f1ca691aadc9841f9111bd9a7ca32550a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)
. .


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR