2-Amino-5-bromo-3-nitropyridine ≥97.0%

Supplier: TCI America

Synonyms: 5-Bromo-3-nitropyridin-2-amine, 5-Bromo-3-nitro-2-pyridylamine

A1556-5G A1556-25G
TCA1556-5GEA 71.1 CAD
TCA1556-5G TCA1556-25G
2-Amino-5-bromo-3-nitropyridine ≥97.0%
2-Amino-5-bromo-3-nitropyridine

CAS Number: 6945-68-2
MDL Number: MFCD00047441
Molecular Formula: C5H4BrN3O2
Molecular Weight: 218.01
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 209

Formula: C₅H₄BrN₃O₂
MW: 218.01 g/mol
Melting Pt: 209 °C
MDL Number: MFCD00047441
CAS Number: 6945-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76332cc30e8182328521d614b6df9806d2b28bfc&vid=76332cc30e8182328521d614b6df9806d2b28bfc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR